On the Home page, you can pin applications to the left pane for easy access from any Ardia platform page. The Data Explorer, Data Viewer, and Instruments applications, as well as the Home page, are pinned to the left pane by default.
Procedure
  1. On the Ardia platform Home page, right-click an application card or select on the application card.
  2. Select Pin to Left Panel.
  3. The application is pinned to the left pane and is accessible from every Ardia platform page.
TIP
You can also pin an application from its page. See Pin an application to the left pane from an application page.