The Thermo Scientific Ardia Platform is composed of the Ardia Core software and Core applications. It is an integrated application and data platform that facilitates the storage, management, and sharing of data between chromatography and mass spectrometry instruments, applications, and the wider ecosystem.
The Ardia core applications and utilities can interact with each other and share capabilities, such as the user interface, identity, notifications, and auditing. The Ardia workspace is accessible via a web browser, which allows users to access their data conveniently and securely.
Additionally, the Ardia platform supports Ardia-ready external software, such as the Thermo Scientific Proteome Discoverer™ software, Thermo Scientific BioPharma Finder™ software, Thermo Scientific Xcalibur™ software, and the Thermo Scientific Chromeleon™ chromatography data system (CDS), which can be connected to the platform.
The following table lists the Ardia Core applications and utilities, as well as the actions you can perform with each one.
Application | Function |
---|---|
Core applications | |
Browse and manage folders, files, and data objects that are stored on the Ardia platform. | |
Visualize and qualitatively analyze chromatography and mass spectrometry data. | |
Manage, schedule, and monitor data acquisition on all instruments connected to the Ardia platform. | |
Utilities | |
Display audit entries generated in the Ardia platform such as user access, data operations, and setting changes. | |
Create and manage application registration codes that are utilized to connect Thermo Scientific desktop applications to the Ardia platform. | |
Define administrative settings that are applied across the Ardia platform. | |
Configure, edit, and remove the identity providers used by the Ardia platform. | |
Add new users and manage existing users in the Ardia platform. Define roles, users, and configure privileges. |