In the Audit Trail Viewer application, you can display audited events, such as user access and data operations.
Prerequisites
- Your role must have the required Ardia Core permission to open the Audit Trail Viewer application. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Do one of the following:
- On the Home page, select the Audit Trail Viewer card under Utilities.
- In the left pane of the Ardia workspace (if pinned), select Audit Trail Viewer.
NOTE
To open an application from the left pane, you must first pin it to the left pane. For more information, see Pin an application to the left pane from the Home page.
- The Audit Trail Viewer application opens. It displays all audit trail types that were generated over the previous two weeks. See Audit Trail Viewer parameters.
- (Optional) Limit your view to specific entry types by selecting one of the following options in the Audit Trail Type list:
- Authentication
- Data Applications
- Global Settings
- External Connections
- User Management
NOTE
If you select All for the Audit Trail Type, it returns entries from all of the Audit Trail types. This could include a large number of results.