In the Audit Trail Viewer application, you can load and run any query in repositories that you have access to.
Procedure
- Open the Audit Trail Viewer.
- Select Load Query.
- The Open dialog box opens.
- Under the Ardia Repository node, locate the folder that contains the query that you want to load.
- Select the query that you want to load.
TIP
If your folder contains many queries, use the search functionality to narrow the results list.
- Select Open.
NOTE
The Open button is available only when you select an Audit-query with the Live status.
- The application loads the selected query and displays the parameter rules on the main Audit Viewer page. It does not automatically run the query.
- Select Run Query to display the results of this query.