In the Audit Trail Viewer application, you can export query results to a PDF file.
NOTE
If you do not create a query, the default query, which includes records from the previous two weeks, is exported to the PDF file.
Procedure
  1. Select Export in the upper-right corner of the results list.
  2. The Confirm Export dialog box opens.
  3. In the Confirm Export dialog box, do one of the following:
  4. To generate a PDF that contains the most recent 5,000 records, select Export.
  5. To generate a PDF that contains a different number of the most recent records, enter the number of records that you want to export in the text box, and then select Export.
  6. The Audit Trailer Viewer generates the PDF and saves the AuditRecords.pdf file to your Downloads folder.
  7. Sample query results PDF
    Sample query results PDF