In the Audit Trail Viewer application, you can create queries to filter the precise events that you want to view.
Procedure
  1. Open the Audit Trail Viewer.
  2. Select Show Query (on the far right of the window).
  3. The application displays a combination of filters as well as And and Or operators to narrow down the search result.
  4. Define the rules in your query as follows:
  • Select And if you want results that meet multiple conditions (returns fewer results).
  • Select Or if you want results that meet any one of a list of conditions (returns more results).
  • Select the condition that you want to search for, such as Object Type, Time Stamp, Username, and so on.
  • Select an operator.
  • Select a target.
  1. Repeat step 3 for each rule in your query.
  2. To add an additional rule to the query, select next to either the Or button or the text box.
  3. (Optional) To add an additional group to the query, select and choose Add Group.
  4. The Audit Viewer adds a separate group where you can create additional sets of rules.
  5. TIP
    Perform step 6 to create both an And rule and an Or rule.
  6. Select Run Query to test your query.
  7. NOTE
    If there are errors in the query, such as an empty value, an error message reports the issue (in the upper-right corner of the window).
Sample query results
Sample query results