The Audit Trail Viewer displays the total number of results and the number of results per page in the lower-left corner of the page. The Audit Trail Viewer displays 100 results per page by default.
The Audit Trail Viewer displays the number of pages of results in the lower-right corner of the page. You can click a page number or use the left and right arrows to navigate between pages.
Procedure
- Select the number of results that you want displayed per page from the Results Per Page dropdown list.
- The Audit Trail Viewer immediately loads the specified number of results.