The Data Explorer application lets you manage the data that is stored in the Ardia Repository.
Prerequisites
  1. Your role must have the required Ardia Core permission to open the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
Procedure
  1. Do one of the following:
  2. On the Ardia Home page, select the Data Explorer card under Core applications.
  3. In the left pane of any Ardia page, select Data Explorer.
  4. NOTE
    To see an application in the left pane, you must first pin the application to the left pane. For more information see Pin an application to the left pane from an application page and Pin an application to the left pane from the Home page.
  5. On a browser window, enter the application URL (Example format: https://domainname.com/app/data-explorer).
  6. The Data Explorer application opens and displays the Ardia Repository, where you can explore and manage your data.
  7. Ardia Repository
    Ardia Repository