In the Data Explorer application, you can view the details of a folder, sequence, injection, document, composite document, synchronized folder, or synchronized sequence.
Prerequisites
- Your role must have the required Ardia Core permission to open the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- You must have the Can View permission (or a higher permission) for the folder to view the details of its contents. For more information, see Manage folder access.
Procedure
- Open the Data Explorer application.
- In the content area, do one of the following:
- Select an item and then select Details on the command bar.
- Select on an item's row and then select Details.
- Right-click an item's row and then select Details.
NOTE
You can only view the Details of one folder, sequence, or file at a time. If you select more than one item, the Details command is disabled.
- The Summary side pane opens and displays the details for the selected object.
- Select in the upper-right corner to close the Summary side pane.