In the Client Registration and Management application, you can restore a disabled registration code.
Prerequisites
  1. Your role must have the required Ardia Core permission to generate an activation code for a client. For more information, see Edit Ardia Core privileges for a role.
Procedure
  1. Open the Client Registration and Management application.
  2. Select all disabled registration codes that you want to restore.
  3. NOTE
    The Enable Codes feature is available only for registration codes that have the Disabled status.
  4. Select Enable codes.
  5. At the prompt, select Enable App(s).
  6. The Add a Comment dialog box appears.
  7. Enter your comment and select Save.
  8. NOTE
    The Add a Comment dialog box appears only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia platform.
    The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.
  9. The application confirms that the selected registration codes are now enabled and updates the status in the table.