Each application associated with the Ardia platform defines its own set of privileges. In the Roles page of the Users and Roles application, you can edit the application privileges assigned to each role.
Prerequisites
- Your role must have the required Ardia Core permission to modify privileges in the Users and Roles application. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Open the Roles page.
- Select a role for which you want to modify privileges, for example: administrator.
- Select Modify Privileges.
- A window opens with the Ardia Core tab selected.
- Select an application tab, for example: the Instruments tab.
- The application displays the assigned privileges for the selected role.
- Turn on the toggles for each privilege to either enable or disable the privilege. For more information, see Edit Instruments user privileges for a role.
- Select Save.
NOTE
If the Save button is disabled, then you do not have the permission to modify privileges.
- The Add a Comment dialog box appears.
- Enter your comment.
NOTE
The Add a Comment dialog box appears only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- The Ardia platform makes the specified application privileges available for all users assigned to that role.