Connected and supporting applications
The Ardia Platform helps bring together a growing list of desktop applications and tools to accelerate data analysis and centralize scientific data and results. Thermo Fisher Scientific recommends using the latest version of these applications to ensure optimal compatibility, functionality, and support. The table below indicates the limitations that may exist with older versions of the desktop applications.
How to interpret the table
  • Full indicates the application is compatible and fully functional with Ardia Core 1.2.
  • Limited indicates expected compatibility, but with limited functionality.
  • [#] indicates a note that provides additional information.
  • [S] indicates a supported version of the connected software with Ardia Core 1.2.
If a software version is not indicated, then it is not supported, and may not be compatible with Ardia Core 1.2 in connected network deployment.
Application
Version
Ardia Core 1.2 Functionality
Ardia Data Sync
1.2 [S]
Full
Ardia Platform Link
1.2 [S]
Full
Compound Discoverer
3.5 [S]
Full
3.4 [S]
Full
Proteome Discoverer
3.4 [S]
Full
3.3 [S]
Full
3.2 [S]
Full
BioPharma Finder
5.4 [S]
Full
5.3 [S]
Full
Chromeleon CDS
7.4 [S]
Full
7.3.2 MUf [S]
Full
7.3.2 MUe [S]
Full
7.3.2 MUd [S]
Full
Xcalibur
4.8 [S]
Full
4.7 SP1 [S]
Full
Skyline™ from MacCoss Lab Software
Daily [S]
Full [1]
  • [1] Customers who contact Thermo Fisher with Skyline issues will be directed to Skyline support. Ardia Platform issues not related to the Skyline API will still go through Thermo Fisher Software Support.