Instrument driver compatibility
Ardia Instruments 1.1.1 and Ardia Data Insights 1.1 applications require communication with instrument hardware to provide full functionality. The drivers listed below are fully compatible with these applications. While newer drivers are expected to be compatible, users are advised to consult with Thermo Fisher Software Support for the most current compatibility information.
  1. Older Instrument Drivers:
    • If an older instrument driver is not listed, it is not supported and may not be compatible with Ardia Core 1.2.
  2. Automated Data Upload and Acquisition Software Compatibility:
    • Automated data upload to Ardia requires compatibility between the instrument and the acquisition software. If the acquisition software is compatible with Ardia, the upload will proceed. Refer to the table above for compatibility between acquisition software and Ardia Core Software 1.2.
    • For the most current compatibility information with acquisition software and individual driver releases, consult with Thermo Fisher Software Support.
Instrument
Driver Version
Orbitrap Astral [1]
2.0
2.0 SP1
2.1
Stellar
1.0 QF1
1.1
Exactive Plus
Exactive Plus EMR
Q Exactive
Q Exactive Focus
Q Exactive Plus
Q Exactive HF
Q Exactive HF-X
Q Exactive UHMR
Q Exactive GC
Exactive GC
Exactive Series 2.13 SP1
Orbitrap Exploris 120
Orbitrap Exploris 240
Orbitrap Exploris 480
Orbitrap Exploris MX
Orbitrap Exploris GC
Orbitrap Exploris GC 240
4.4*
5.0 +
5.1 +
Orbitrap Fusion
Orbitrap Fusion Lumos
Orbitrap Ascend
Orbitrap Ascend Editions
Orbitrap Eclipse
Orbitrap ID-X
4.2 QF1 with patch*
4.3 +
TSQ Quantis
TSQ Altis
TSQ Fortis
TSQ Quantiva
TSQ Endura
TSQ Quantis Plus
TSQ Altis Plus
TSQ Fortis Plus
3.4 SP2 QF1
4.0 QF1
5.0
LC Devices
3.2 SP3
Aria MX
2.8
SII for Xcalibur
1.7
1.8
1.9
Vanquish Core LC
Vanquish Horizon LC
Vanquish Flex LC
Ultimate 3000
Installed with Chromelon or SII for Xcalibur
LC Vanquish Neo LC
1.6
ICS-4000 IC
ICS-6000 IC
Acquion IC
Integrion IC
ASE
Easion
Installed with Chromelon or SII for Xcalibur
AI/AS 1310 Autosampler
TriPlus RSH/RSH Smart Autosampler
Triplus 500 Headspace Autosampler
Triplus 100 LS Autosampler
Trace 1300 Series GC
Trace Ultra GC
Installed with Chromelon or SII for Xcalibur
AI/AS 1610 Autosampler
Trace 1600 Series GC
1.1
TSQ 9000/9610 Triple Quadrupole
ISQ 7000/7610 Single Quadrupole
ISQ EC Single Quadrupole
ISQ EM Single Quadrupole
5.1
5.2
Evosep One
Evosep Eno
3.0.18.4
1.0.98
Waters Acquity LC
3.0.0.34
Agilent LC
3.0.22.0
Shimadzu LC and GC
3.2.1.0
* Instrument/driver combination allows for the autonomous creation of instrument hardware service tickets feature
[1] Not supported in Chromeleon CDS at this time