Ardia server compatibility and upgrade table
This table defines the supported upgrade scenarios and the validated system baseline at the time of release; component revisions are governed by release notes unless compatibility changes.
|
Existing OS |
Existing Platform |
Existing Firmware |
Target OS |
Target Platform |
Target Firmware |
Upgrade Category |
Notes |
|---|---|---|---|---|---|---|---|
|
Ardia Server OS 2.1.0(Ubuntu 20.04.6) |
Ardia Platform 1.1 / 1.1.1 |
T550_BOOTABLE_24.05.31 (minimum recommended) |
Ardia Server OS 3.0.7(Debian 12.12) |
Ardia Platform 1.2.0.1 |
T550_BOOTABLE_25.09.29 |
Full Upgrade |
Only Ardia Core Software versions 1.1 and 1.1.1 are supported as upgrade starting points for this release |
|
Ardia Server OS 3.0.7(Debian 12.12) |
Ardia Platform 1.2.0.1 |
T550_BOOTABLE_25.09.29 (minimum recommended) |
Future compatibility to be defined |
|
APC Version |
Supported Ardia Server OS |
Supported Ardia Platform |
|---|---|---|
|
APC 1.0.9 |
Ardia Server OS 3.0.7 (Debian 12.12) |
Ardia Platform 1.2.0.1 |