In the Global Settings utility, you can define the settings for the Ardia Instruments application.
Prerequisites
- Your role must have the required Ardia Core permission to open the Global Settings utility. For more information, refer to "Edit Ardia Core privileges for a role" in the Ardia Core User Guide.
Procedure
- Select the Instruments tab.
- The Modify Instruments Application Settings page opens, where you can configure the Scheduler settings.
- Select the options for all required settings parameters. For more information on the parameters, see Instruments application settings parameters.
- Select Save.
NOTE
To avoid losing any unsaved changes, save all changes before you navigate away from the Global Settings page.
- The Add a Comment dialog box opens.
- Enter your comment and select Save.
NOTE
The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- A message appears confirming the successful update.