You can temporarily disconnect the Proteome Discoverer application from the Ardia platform by switching to local mode.
Procedure
  1. On the Proteome Discoverer menu bar, select Ardia Server > Sign Out.
  2. The application disconnects from the Ardia Platform.
  3. In the bottom-right corner, the application displays the status of its connection to Ardia.