You can temporarily disconnect the BioPharma Finder application from the Ardia platform by switching to local mode.
Procedure
- On the BioPharma Finder home page, select Help > Ardia Platform Management > Disconnect.
Figure Disconnect option - Select OK.
- The application restarts in local mode.