You can deregister the BioPharma Finder application from the Ardia Platform.
Prerequisites
- Ensure all data processing is complete and all files are closed.
Procedure
- On the BioPharma Finder title bar, select Help > Ardia Platform Management > Deregister with the Ardia Platform.
- The software restarts in local mode.