You can deregister the Xcalibur data system from the Ardia Platform.
Prerequisites
- Close the Xcalibur data system.
Procedure
- Open Microsoft™ Services.
- Locate and right-click Thermo Xcalibur Service in the right pane.
Figure Thermo Xcalibur Service - Select Stop.
- Open the File Explorer.
- Navigate to the C:\ProgramData\Thermo\Xcalibur\HyperBridge folder.
NOTE
If the HyperBridge folder is hidden, unhide it before proceeding.
Figure HyperBridge folder - In the HyperBridge folder, rename the ArdiaDataStorage.db file as follows: ArdiaDataStorage.db.unregister.
- In the Services window, right-click Thermo Xcalibur Service, and then select Start.
- The Xcalibur data system is deregistered from the Ardia Platform.