To deregister the Chromeleon CDS from the Ardia Platform, you must modify the global policies in the Chromeleon Administration Console and remove the respective registration code from the Client Registration and Management utility.
Prerequisites
- Ensure there are no files open in the Chromeleon console.
Procedure
- Open the Chromeleon Administration Console and sign in.
- In the left pane, select Global Policies.
- Under List of policies, select User Mode.
Figure Enable Adia Integration - Clear the Enable Ardia Integration checkbox.
- Select Save Policies.
- Open the Ardia Platform.
- Under Utilities, select the Client Registration and Management card.
- Select the row that has Chromeleon Administration Console under App Name and that has the IP address of the computer where the Chromeleon CDS is installed under PC Name.
- Select Remove Apps.
- Restart the Chromeleon CDS.