You can temporarily disconnect the Xcalibur data system from the Ardia Platform by switching to local mode.
Procedure
  1. In the Xcalibur Roadmap View, select Ardia Platform.
  2. The Thermo Scientific Ardia Platform home page opens.
  3. Select the User icon in the top-right corner.
  4. Select Sign Out.
  5. The Xcalibur application is disconnected from the Ardia Platform and the Ardia functionalities are disabled.