You can register the Xcalibur application to the Ardia Platform using a registration code. This registration code can be generated in the Ardia Client Registration and Management utility.
Prerequisites
- Xcalibur 4.7 or later version is installed on the computer.
Procedure
- In the Client Registration and Management utility, generate a registration code.
- Copy the registration information.
- Open the Xcalibur data system.
- In the Roadmap view, select Ardia Platform.
Figure Roadmap view - The Thermo Scientific Ardia Platform home page opens.
- On the home page, the Xcalibur Registration card is enabled, and the Data Explorer and the Acquisition and Upload Status cards are disabled.
Figure Xcalibur Registration card enabled - On the Xcalibur Registration card, select Open.
- The Ardia Platform Registration dialog box opens.
- Paste the registration information that you copied from the Client Registration and Management application into the Platform URL box and the Registration Code box.
- Select Continue.
- A message confirming the successful registration is displayed.
- Select Close.
- The Thermo Scientific Ardia Platform page displays a connection icon
in the top-right corner. This icon indicates that the application is registered but not connected.
- A browser window opens prompting you to sign in to the Ardia Platform.
- Sign in to the Ardia Platform.
- A message confirming that the application is connected to the Ardia Platform is displayed.
- Close the browser window.
- On the Xcaliber Thermo Scientific Ardia Platform page, the account name and its associated access level are displayed in the top-right corner.
- The connection icon turns green
.
- The Data Explorer card and Acquisition and Upload Status card are enabled.