To connect the Chromeleon CDS to the Ardia Platform, you must add the Chromeleon User Management information in the Ardia Installer.
Prerequisites
- Download the Thermo.Ardia.Server.Installer.latest version.exe file from the Product Downloads portal.
Procedure
- Open the Thermo.Ardia.Server.Installer.latest version.exe file to execute the Ardia Installer.
- Select
Configure.
- The Configure dialog box opens.
- Select the Override File tab.
- The Override File page opens with the Identity Config pane displayed by default.
- In the Identity Config pane, enter the Chromeleon User Management information as follows:
- Enter the Fully Qualified Domain Name (FQDN) of the server hosting the Chromeleon domain controller in the cmUserManagement.host box.
- Enter 1325 in the cmUserManagement.port box.
- Select Apply.
- Select
Install.
- The installer overrides the Identity Configuration information in the Ardia configuration file.
- Close the Installer.