You must enable a data vault to be used with the Ardia Platform. To connect an existing data vault to the Ardia Platform, you must edit the data vault configuration. You can also Create a new data vault and connect it to the Ardia Platform.
Procedure
  1. On the computer that contains your target data vault, open the Chromeleon Administration Console.
  2. Select Local Machine in the left pane.
  3. Select Manage Data Vaults.
  4. The Data Vault Manager dialog box opens.
  5. Under Local Data Vaults, select the data vault that you want to enable.
  6. Select Configure.
  7. The Configure Data Vault dialog box opens.
  8. Select the Automatic Upload of Sequences & Injection Data to Ardia checkbox.
  9. Select OK and then close the dialog box.