You must enable a data vault to be used with the Ardia Platform. If you are unable or do not want to connect an existing data vault to the Ardia Platform, you can create a new data vault and connect it to the Ardia Platform.
Procedure
  1. Open the Chromeleon Administration Console.
  2. Select Local Machine in the left pane.
  3. Select Manage Data Vaults.
  4. The Data Vault Manager dialog box opens.
  5. Select Create.
  6. The Choose Data Vault Type page opens.
  7. Select the type of data vault that you want to create.
  8. For more information on the Data Vault types, refer to the Chromeleon Online Help.
  9. Select Next.
  10. Enter a name for the data vault in the Name box.
  11. Select the Automatic Upload of Sequences & Injection Data to Ardia checkbox.
  12. Select Finish.
  13. Close the Data Vault Manager dialog box.
  14. Open the Chromeleon main console.
  15. The Ardia Login dialog box opens.
  16. Sign in to the Ardia Platform using your Chromeleon login credentials.