To connect the Chromeleon CDS to the Ardia Platform, you must modify the Global Policies.
Procedure
- Open and sign in to the Chromeleon Administration Console.
- In the left pane, select Global Policies.
Figure Chromeleon Administration Console - Under List of Policies, select User Mode.
- Select the Enable Ardia Integration checkbox.
- The Ardia Registration dialog box opens.
Figure Enter the Ardia Platform URL - Enter the Ardia Platform URL, and then select Connect.
- The dialog box displays a hyperlink, a numeric code, and a QR code.
Figure Code for connecting to Ardia Platform - Copy the numeric code and then select the hyperlink.
- The Ardia Identity page opens in a browser.
- (Optional) If prompted, sign in to the Ardia Platform.
- Enter the copied code in the User Code box and then select Submit.
Figure Enter the code - If the connection is established, a success message is displayed.
- In the Chromeleon Administration Console, the Change Client Registration button, the Ardia URL, and the Enable Single Sign-On option are available.
Figure New options in the Chromeleon Administration Console - Select Save Policies at the top of the Chromeleon Administration Console's right pane.
- A dialog box notifies you that the Administration Console requires a restart to perform access controlled operations.
- Select OK.
- Close the Administration Console and reopen it.
- The Ardia Login dialog box opens.
- Sign in to the Ardia Platform using your Chromeleon login credentials.