In the Audit Trail Viewer utility, you can revert an object to a previous version.
Prerequisites
- Your role must have the required Ardia Core permissions to revert an object to a previous version. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Open the Audit Trail Viewer.
- View the complete audit trail of the object by doing one of the following:
- Select a checkbox corresponding to an audit entry you want to view (on the left side of the table), then select View Audit Trail that displays above the table.
- Right-click anywhere in an entry row and select the View Audit Trail option.
- Select the checkbox for the single object version you want to revert to.
NOTE
You cannot revert to the current version of the object.
- The Revert button displays above the table and is enabled.
- Select Revert.
- The object is restored as it was in the selected version.
- A new version of the object is added to the table with the appropriate timestamp and it is set as the current version.