The following table lists the data types that are compatible with the Ardia Repository and the icons that represent each data type.
Data type icon | Description | Integration type |
|---|---|---|
Represents a folder. | NA | |
Represents a sequence such as:
| Integral | |
Represents an injection. | Integral | |
Represents a PDF file. | Supported | |
Represents an image, including the following formats:
| Supported | |
Represents PD result files, including the following:
| Integral | |
Represents CD result files. | Integral | |
Represents other data types. These types are categorized based on the data's source and are listed here. | ||
Ardia Platform | ||
Audit Query | Integral | |
Index | Integral | |
BioPharma Finder | ||
BPF method | Integral | |
BPF intact report | Integral | |
BPF intact report template | Integral | |
BPF intact report workbook | Integral | |
BPF intact experiment | Integral | |
BPF intact workbook | Integral | |
BPF oligo report | Integral | |
BPF oligo report template | Integral | |
BPF oligo report workbook | Integral | |
BPF oligonucleotide experiment | Integral | |
BPF oligonucleotide workbook | Integral | |
BPF peptide report | Integral | |
BPF peptide report template | Integral | |
BPF peptide report workbook | Integral | |
BPF peptide experiment | Integral | |
BPF peptide workbook | Integral | |
BPF result | Integral | |
BPF topdown experiment | Integral | |
BPF workbook | Integral | |
Chromeleon CDS | ||
Chromeleon 7 data | Integral | |
Compound Discoverer | ||
CD result data | Integral | |
CD result details | Integral | |
CD result details view | Integral | |
CD result metadata | Integral | |
CD result view | Integral | |
CD result | Integral | |
CD study | Integral | |
Generic files | ||
.csv | Supported | |
mzIdentML | Supported | |
Open proteomics identification | Supported | |
Protein Data Bank | Supported | |
.rft | Supported | |
.txt | Supported | |
.xml | Supported | |
Instrument data | ||
Mascot generic format | Supported | |
Open, XML-based format for mass spectrometry data | Supported | |
Open, XML-based format for mass spectrometry data | Supported | |
Open, XML-based format for mass spectrometry proteomics data | Supported | |
Thermo Scientific raw data | Integral | |
Microsoft™ Office | ||
Microsoft™ Word document | Supported | |
Microsoft PowerPoint™ document | Supported | |
Microsoft Excel™ document | Supported | |
ProSightPD | ||
ProSight Lite result | Integral | |
ProSight database | Integral | |
ProSight result | Integral | |
TDValidator result | Integral | |
Proteome Discoverer | ||
CHIMERYS inclusion list | Integral | |
PD quantification method | Integral | |
PD result data (processing) | Integral | |
PD processing result details | Integral | |
PD processing result details view | Integral | |
PD processing result metadata | Integral | |
PD processing result view | Integral | |
PD result data (consensus) | Integral | |
PD consensus result details | Integral | |
PD consensus result details view | Integral | |
PD consensus result metadata | Integral | |
PD consensus result view | Integral | |
PD study | Integral | |
TraceFinder | ||
TraceFinder compound database | Supported | |