In the Data Explorer application, you can revert a file to a previous version.
Prerequisites
- Your role must have the required Ardia Core permissions to revert a file to a previous version in the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- You must have the Can Edit permission (or a higher permission) for the folder to revert a file in it to a previous version. For more information, see Manage folder access.
Procedure
- Open the Data Explorer application.
- Navigate to the folder that contains the file that you want to revert to a previous version.
- Open the Versions pane.
- In the Versions pane, select the checkbox of the version to which you want to revert the file.
- The Revert command becomes available on the Versions pane command bar.
NOTE
If you select more than one version or the current version, the Revert command is unavailable.
Figure Revert a file version - Select the Revert command.
- The content of the file is restored to how it was in the selected version.
- In the Versions pane, a new version is added with a date-time stamp and it is set as the Current version.