You can view the complete audit trail of an item in the Ardia Repository using the Data Explorer.
Prerequisites
- Your role must have the required Ardia Core permission to open the Audit Trail Viewer utility. For more information, see Edit Ardia Core privileges for a role.
- Your role must have the required Ardia Core permission to open the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- Your role must have the Can View permission (or a higher permission) for the folder to view its content. For more information, see Manage folder access.
Procedure
- Open the Data Explorer application.
- The Data Explorer application opens and displays the Ardia Repository.
- In the content area, do one of the following:
- Select a checkbox corresponding to an item and then select View Audit Trail from the command bar.
- Right-click anywhere in an item row and select the View Audit Trail option.
- The Audit Trail Viewer opens in a new tab and displays all audit entries related to the selected item. For more information, see View the complete audit trail of an object.