In the Audit Trail Viewer utility, you can view the complete audit trail of an object that triggered the audit record.
Procedure
- Open the Audit Trail Viewer.
- Do one of the following:
- Select a checkbox corresponding to an audit entry you want to view (on the left side of the table), then select View Audit Trail that displays above the table.
- Right-click anywhere in an entry row and select the View Audit Trail option.
- The Audit Trail Viewer opens in a new tab and displays all entries related to the selected object. The entry that was previously selected is highlighted and the filtering and querying options are no longer displayed.