You can customize the columns in the Audit Trail Viewer to display only the ones you want to view and to reorder the columns.
Procedure
- Select
.
- The Edit Columns side pane opens.
- To show or hide one or more columns, do one of the following:
- Select the checkbox for a column name to show that column.
- Clear the checkbox for a column name to hide that column.
- To reorder the columns, drag the column name to a new position.
- Select Restore Defaults to set the column visibility, order, and width back to the original, default settings.
- Do one of the following:
- Select Apply to proceed with the changes.
- Select Cancel to close the dialog box without applying the changes.