You can adjust the column width in the Audit Trail Viewer utility to fit your requirements.
Procedure
  1. Point to the right side of a column boundary until the resize cursor appears.
  2. Drag the column boundary to adjust the width to your preference.
NOTE
To set the column width back to the original, default settings, select Restore Defaults in the Edit Columns side pane.