With the Audit Trail Viewer utility, you can view a log of user activity within the Ardia Platform. The log is displayed as a table with the following columns: Timestamp, Object Type, Object Name, Version, Application, Action, Username, Role, Comment, Path, and Additional Info.
The following table defines the columns that appear in the Audit Trail Viewer utility.
Column | Description |
|---|---|
Timestamp | Specifies the time at which an audited event occurred. |
Object Type | Specifies the type of object that triggered the audit record. Examples: Authenticated User Session, Folder, Injection, Sequence, Raw File, and so on. |
Object Name | Specifies the name of the object that triggered the audit record. |
Version | Specifies the version of the object that triggered the audit record. |
Application | Specifies the name of the application in which the audit event occurred. |
Action | Specifies the type of activity that was performed and recorded in the audit log. |
Username | Specifies the name of the user who performed the audited event. |
Role | Specifies the role that the user was logged in as at the time the audited event was performed. |
Comment | Specifies user-added comments (if the Prompt for Comments When Saving toggle is turned on in the Global Settings application and the user has provided a comment). |
Path | Specifies the Ardia Repository file path of the object that triggered the audit record. |
Additional Info | Captures any system-generated additional information, if available. |