An Ardia administrator can assign roles, users, and privileges using the Users and Roles utility.
The first user comes preconfigured with the default Ardia Platform installation. This user is automatically granted the super administrator role, which allows them to access the full scope of the Ardia Platform.
When a new role is created, it does not have any privileges enabled by default. An administrator can upgrade the role as needed.
Term | Description |
|---|---|
Refers to a set of privileges that defines the actions that a user with that role can perform within the Ardia Platform. Roles are assigned based on user responsibilities and job functions. | |
Refers to an individual who is assigned a specific role within the Ardia Platform. This role determines the user's level of access and privileges within the platform. | |
Refer to the actions or functionality that a user with a specific role can perform within the Ardia Platform. Each role has its own set of privileges, which can be assigned or removed as needed by an administrator. The Ardia Platform has two sets of privileges:
|