If you are experiencing issues with the Ardia Platform, see the following table to help you identify and troubleshoot problems. If you are unable to resolve a problem using the information provided, contact your administrator or the Thermo Support team.
Problem | Possible cause | Solution |
|---|---|---|
If you use Microsoft Entra ID as your identity provider, you might encounter a situation where you cannot sign in, even after the previous user has signed out of the Ardia Platform. | The previous user is signed out of the Ardia Platform but the user's Microsoft account is not completely signed out. | To completely sign out of Microsoft Entra ID, follow these steps: Procedure
|