Using the Data Explorer application, you can upload raw data file as a new sequence in the Ardia Repository.
Prerequisites
- Your role must have the required Ardia Core permission to upload data in the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- You must have the Can Edit permission for the folder to which you want to upload the files. For more information, see Manage folder access.
Procedure
- Open the Data Explorer application.
- Navigate to the folder where you want to upload a raw data file.
- Select Upload Files.
NOTE
The Upload Files button is available only for folders with one of these statuses: Live, Live Archive Locked, or Live Archive Incomplete.
- The Open dialog box opens.
- Select one or more raw data files and then select Open.
TIP
You can also drag the raw data files directly from the Windows File Explorer to the Data Explorer page.
- The Give New Sequence a Name dialog box opens.
- The Sequence name is pre-filled with the name of the first raw data file selected.
- The dialog box lists all the raw data files selected.
- (Optional) Edit the sequence name.
- (Optional) Select
next to the raw data files that you want to remove from the selection.
- Select Upload.
- The Add a Comment dialog box opens.
NOTE
You can upload additional files during an ongoing upload.
- Enter your comment and then select Save.
NOTE
The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- A progress bar informing the status of the ongoing upload is displayed.
NOTE
At this stage, you can cancel the entire upload or an individual file upload. For more information, see Cancel an upload.
- When the upload is complete, the progress bar closes and the system sends an in-app notification confirming that the files are uploaded.
- A new sequence with the uploaded raw data files is added to the Ardia Repository.