Archiving is the process of moving data from the Ardia server to a specified location that is defined during the Ardia Platform setup. Archiving is enabled only if the archive path is specified during the setup process. In the Data Explorer application, you can archive folders, sequences, or documents.
The Archive command is available when you select one or more items with specific statuses, as shown in the following table.
Item | Status | Archive command |
|---|---|---|
| Available | |
Unavailable | ||
| Any status | Unavailable |
Prerequisites
- Your role must have the required Ardia Core permission to archive data in the Data Explorer application. For more information, see Edit Ardia Core privileges for a role.
- You must have Can Edit permission (or a higher permission) for a folder to archive the folder or its content (documents or sequences). For more information, see Manage folder access.
- Archiving must be enabled during the Ardia Server deployment. For more information, see Enable backup and archive storage during the Ardia Server deployment.
Procedure
- Open the Data Explorer application.
- Navigate to the folder that contains the item that you want to archive.
- In the content area, do one of the following:
- Select
on the item's row and then select Archive.
- Right-click the item's row and then select Archive.
- Select the checkboxes of one or more items and then select Archive on the command bar.
TIP
To select consecutive files, press and hold the SHIFT key and select the first row and the last row of the items that you want to select.
To select non-consecutive files, press and hold the CTRL key and select each row.
To select all the files in the folder, select the checkbox next to the Name column heading.
TIP
You can select and archive a combination of folders, sequences, and documents together.
- The availability of the Archive command depends on the item types and the status.
- In the Archive Confirmation dialog box, select Archive.
- The Add a Comment dialog box opens.
- Enter your comment and then select Save.
NOTE
The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- Whenever you save any changes, the Ardia Platform sends an in-app notification that indicates the progress and success of the archiving process.
- When an item is successfully archived, its status changes from Live to Archived. On archival failure, the status changes from Live to Live Archive Incomplete.