You can access notifications that were previously cleared.
NOTE
Cleared notifications are automatically deleted after a specific number of days. Users with appropriate administrative rights can set this duration in the Global Settings utility (see Ardia Core settings parameters).
Procedure
  1. Open the Notifications pane, then select View Cleared Notifications at the bottom.
  2. A list of cleared notifications opens.
  3. To return to the main notification list, select in the upper-right corner of the navigation pane.