You can filter notifications based on the following criteria:
- Severity: Error, Warning, Info, or Success
- State: Read or Unread
Procedure
- Open the Notifications pane, then select Filter in the upper-right corner.
- The notification filter bar opens.
NOTE
The Filter command is disabled when there are no active notifications.
- To choose the filter criteria, do the following as needed:
- In the Severity list, select Error, Warning, Info, or Success.
- In the State list, select Read or Unread.
- Depending on your selection, the Notifications pane displays the filtered notifications.