In the Audit Trail Viewer utility, you can create and save custom queries.
Prerequisites
  1. You have created a query in the Audit Trail Viewer (see Create a new query).
Procedure
  1. When you are satisfied with your query (and have tested it), select Save Query.
  2. The Save As dialog box open.
  3. Under the Ardia Repository node, select the location to save your query.
  4. TIP
    To create a new folder, select New Folder.
  5. Enter a name for your query or select an existing query in the list to overwrite.
  6. NOTE
    The query name cannot exceed 128 characters including the “.audit-query” filename extension.
  7. (Optional) Enter a description for your query.
  8. Select Save.