The Audit Trail Viewer displays audit entries generated in the Ardia Platform, such as user access, data operations, and setting changes. Audit records include user information, timestamps, user-added comments, and details about the entry (see Audit Trail Viewer parameters). You can create queries to filter the precise events that you want to view, compare versions, and revert an object to a previous version directly in the application.
NOTE
Only users with the appropriate Ardia Core permissions may access the Audit Trail Viewer utility. To request access to the application, contact your administrator.
With the appropriate permissions, you can: