In the Audit Trail Viewer utility, you can load and run any query in repositories that you have access to.
Procedure
  1. Open the Audit Trail Viewer.
  2. Select Load Query.
  3. The Open dialog box opens.
  4. Under the Ardia Repository node, locate the folder that contains the query that you want to load.
  5. Select the query that you want to load.
  6. TIP
    If your folder contains many queries, use the search functionality to narrow the results list.
  7. Select Open.
  8. NOTE
    The Open button is available only when you select an Audit-query with the Live status.
  9. The application loads the selected query and displays the parameter rules on the main Audit Viewer page. It does not automatically run the query.
  10. Select Run Query to display the results of this query.