In the Audit Trail Viewer utility, you can export query results to a PDF file.
NOTE
If you do not create a query, the default query, which includes records from the previous two weeks, is exported to the PDF file.
Procedure
- Select Export in the upper-right corner of the results list.
- The Confirm Export dialog box opens.
- In the Confirm Export dialog box, do one of the following:
- To generate a PDF that contains the most recent 5,000 records, select Export.
- To generate a PDF that contains a different number of the most recent records, enter the number of records that you want to export in the text box, and then select Export.
- The Audit Trailer Viewer generates the PDF and saves the AuditRecords.pdf file to your Downloads folder.
Figure Sample query results PDF