In the Client Registration and Management utility, you can permanently remove a registration code. After deregistration, the application can no longer connect to the Ardia Platform and is removed from the list of registered applications.
Prerequisites
  1. Your role must have the required Ardia Core permission to generate an activation code for a client. For more information, see Edit Ardia Core privileges for a role.
Procedure
  1. Open the Client Registration and Management utility.
  2. Select the applications whose registration codes you want to remove.
  3. NOTE
    You can select multiple applications at once, but do not use the SHIFT or CTRL keys to select multiple codes.
  4. Select Remove Apps.
  5. At the prompt, select Yes.
  6. The Add a Comment dialog box opens.
  7. Enter your comment and select Save.
  8. NOTE
    The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
    The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.
  9. The Ardia Platform immediately removes the selected registration code from the list of registered applications and renders this registration code invalid for future use.
  10. The registered application receives a warning message that this registration code is now void. Applications using this registration code can no longer access the Ardia Platform.