In the Client Registration and Management utility, you can disable a registration code that is used to connect a desktop application to the Ardia Platform. Disabling a registration code temporarily restricts the connection of the application to the Ardia Platform.
NOTE
Disabling a code is different from removing one, as a disabled code can be re-enabled, while removing a code is permanent.
Prerequisites
- Your role must have the required Ardia Core permission to generate an activation code for a client. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Open the Client Registration and Management utility.
- Select all active registration codes that you want to disable.
NOTE
The Disable Apps feature is available only for registration codes that have the Active
status.
- Select Disable Apps.
- At the prompt, select Yes.
- The Add a Comment dialog box opens.
- Enter your comment and select Save.
NOTE
The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- The selected registration codes are now disabled.
- The registered application receives a warning message that this registration code is now disabled. Applications using this registration code can no longer access the Ardia Platform.