In the Client Registration and Management utility, you can create new registration codes. Registration codes are required for client applications to register with the Ardia Platform.
Prerequisites
- Your role must have the required Ardia Core permission to generate an activation code for a client. For more information, see Edit Ardia Core privileges for a role.
Procedure
- Open the Client Registration and Management utility.
- Select Add Registration Code.
- The Add a Comment dialog box opens.
- Enter your comment and select Save.
NOTE
The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.- The new code is added to the list of registration codes.
- Provide this registration code to users so that they can register their application.
- Newly added registration codes are highlighted in green until they are used by the application.