On the Roles page in the Users and Roles utility, you can edit the privileges that are associated with the Data Explorer application, Data Viewer application, and Ardia utilities for each role.
Prerequisites
  1. Your role must have the required Ardia Core permission to modify privileges in the Users and Roles utility.
Procedure
  1. Open the Roles page.
  2. Select a role for which you want to modify privileges, for example: Administrator.
  3. Select Modify Privileges.
  4. The Assign Role Privileges window opens with the Ardia Core tab selected, where you can see the assigned privileges for the selected role.
  5. Turn on the toggles for the privileges that you want to give the role.
  6. NOTE
    You cannot disable both Assign Privileges privilege and Open Users and Roles Application privilege for a role unless another role has both these privileges. At least one role must have both these privileges.
  7. Select Save.
  8. NOTE
    If the Save button is disabled, then you do not have the permission to modify privileges.
  9. The Add a Comment dialog box opens.
  10. Enter your comment.
  11. NOTE
    The Add a Comment dialog box opens only if the Prompt for Comments When Saving toggle is turned on in the Global Settings (according to the audit policy in your organization). See Configure global settings for the Ardia Platform.
    The comment that you enter is added to the Comment column of the Audit Trail Viewer. See Open the Audit Trail Viewer.
  12. The Ardia Platform makes the specified permissions available for all users assigned to that role. For more information on the available Ardia Core permissions, see Ardia Core privileges.