A new revision of the operating system image, Revision 2.1.0, is now provided and installed on all new servers. This revision introduces digitally signed NVIDIA drivers, which allows for Secure Boot to be enabled. Further information about the drivers can be found here. Revision 2.1.0 also removes graphical user interface (GUI) software, including the LXQt desktop and X Server.