A new script, os-upgrade.sh, is now available to patch the operating system (OS) of the Ardia Server. This script updates the OS to a patch baseline that has been tested and validated by Thermo Fisher Scientific. It also supports offline patching, which enables updates for Ardia Servers in isolated or air-gapped environments.